Learn More
Sigma Aldrich Fine Chemicals Biosciences Vitamin K2 MK 7 456788 13C1MG
Catalog No. 502907419
Encompass
Vitamin K2 also known as menaquinone is a polyisoprenoid-substitutednbspnapthoquinone. Menaquinone-7 (MK-7) is a subtype of vitamin K2. It is found in animals soy protein fermented products and is primarily produced by bacteria.Vitamin K2 (MK-7)-4AE 5 6 7 8 8AE-13C6 is a vitamin K2 isotope wherein C-4AE C-5 C-6 C-7 C-8 and C-8AE carbons are replaced by 13C6.
Catalog No. 50-290-7419 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 9000751MG
Vitamin K2 also known as menaquinone is a polyisoprenoid-substitutednbspnapthoquinone. Menaquinone-7 (MK-7) is a subtype of vitamin K2. It is found in animals soy protein fermented products and is primarily produced by bacteria.Vitamin K2 (MK-7)-4AE 5 6 7 8 8AE-13C6 is a vitamin K2 isotope wherein C-4AE C-5 C-6 C-7 C-8 and C-8AE carbons are replaced by 13C6.