Learn More
Sigma Aldrich Fine Chemicals Biosciences Vitamin K2 MK 7 5678 d42 m1MG
Catalog No. 502907872
Encompass
Vitamin K2 also known as menaquinone is a polyisoprenoid-substitutednbspnapthoquinone. Menaquinone-7 (MK-7) is a subtype of vitamin K2. It is found in animals soy protein fermented products and is primarily produced by bacteria.Vitamin K2 (MK-7)-(5 6 7 8-d4 2-methyl-d3) is a deuterated vitamin K2 wherein C-5 C-6 C-7 C-8 and 2-methyl protons are replaced by deuterium.
Catalog No. 50-290-7872 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 9000741MG
Vitamin K2 also known as menaquinone is a polyisoprenoid-substitutednbspnapthoquinone. Menaquinone-7 (MK-7) is a subtype of vitamin K2. It is found in animals soy protein fermented products and is primarily produced by bacteria.Vitamin K2 (MK-7)-(5 6 7 8-d4 2-methyl-d3) is a deuterated vitamin K2 wherein C-5 C-6 C-7 C-8 and 2-methyl protons are replaced by deuterium.