Learn More
Medchemexpress LLC Vitamin K3-d8 | 478171-80-1 | MFCD01317514 | 98.9% | 180.23 g/mol | C11D8O2 | 5 MG
SDP
Catalog No. 5000262906
Encompass_Preferred
Vitamin K3-d8 is a deuterium-labeled analogue of vitamin K3 (menadione) used as an internal standard and tracer for quantitative analytical methods. It is supplied as a small-quantity research reagent for method development and accurate quantification in NMR, GC-MS, and LC-MS workflows.
  • Deuterium-labeled internal standard for vitamin K3
  • Suitable for NMR, GC-MS, and LC-MS quantitative analysis
  • Molecular formula C11D8O2; molecular weight 180.23 g/mol
  • High purity (manufacturer lists ~98.9%)
  • Supplied in small-mass quantities suitable for method development
  • For research use only; not for human or clinical use
Catalog No. 50-002-62906 Supplier Medchemexpress LLC Supplier No. HY132389S5MG
Vitamin K3-d8 is a deuterium-labeled analogue of vitamin K3 (menadione) used as an internal standard and tracer for quantitative analytical methods. It is supplied as a small-quantity research reagent for method development and accurate quantification in NMR, GC-MS, and LC-MS workflows.
  • Deuterium-labeled internal standard for vitamin K3
  • Suitable for NMR, GC-MS, and LC-MS quantitative analysis
  • Molecular formula C11D8O2; molecular weight 180.23 g/mol
  • High purity (manufacturer lists ~98.9%)
  • Supplied in small-mass quantities suitable for method development
  • For research use only; not for human or clinical use