Learn More
Cayman Chemical Vitamin K3-d8, Purity: ≥99% deuterated forms (d1-d8), CAS: 478171-80-1, 50 mg
SDP
Catalog No. 502944258
Encompass_Preferred
An internal standard for the quantification of vitamin K3 by GC- or LC-MS
Catalog No. 50-294-4258 Supplier Cayman Chemical Supplier No. 4128550 MG
An internal standard for the quantification of vitamin K3 by GC- or LC-MS