Learn More
Cayman Chemical Vitamin K3-d8, Purity: ≥99% deuterated forms (d1-d8), CAS: 478171-80-1, 5 mg
SDP
Catalog No. 502978450
Encompass_Preferred
An internal standard for the quantification of vitamin K3 by GC- or LC-MS
Catalog No. 50-297-8450 Supplier Cayman Chemical Supplier No. 412855 MG
An internal standard for the quantification of vitamin K3 by GC- or LC-MS