Learn More
Medchemexpress LLC HY-B1508 200mg Medchemexpress, Vitamin K4 CAS:573-20-6 Purity:>98%
SDP
Catalog No. 501932957
Encompass_Preferred
Medchemexpress, HY-B1508 200mg Vitamin K4 CAS:573-20-6 Vitamin K4 is a chemically synthesized Vitamin K which plays an important role in the normal blood coagulation system. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2957 Supplier Medchemexpress LLC Supplier No. HYB1508200MG
Medchemexpress, HY-B1508 200mg Vitamin K4 CAS:573-20-6 Vitamin K4 is a chemically synthesized Vitamin K which plays an important role in the normal blood coagulation system. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.