Learn More
Medchemexpress LLC Vitamin K4 | 573-20-6 | 99.9% | 1 G
SDP
Catalog No. 5000411182
Encompass_Preferred
Vitamin K4 is a chemically synthesized Vitamin K that plays an important role in the normal blood coagulation system. It arrests cells in the S phase of the cell cycle and induces apoptosis, and can be used for research on cancer, including prostate cancer and osteosarcoma.
  • Inhibits cell proliferation and induces caspase-dependent apoptosis in PC-3 cells.
  • Induces S phase cell cycle arrest in PC-3 and U2OS cells.
  • Inhibits the growth of U2OS cells.
  • Increases ROS generation and disrupts mitochondrial membrane potential in U2OS cells.
  • Inhibits the migration of U2OS cells in vitro.
  • Modulates expression of Bcl-2, Bax, PARP, and cleaved-caspase-3.
Catalog No. 50-004-11182 Supplier Medchemexpress LLC Supplier No. HYB15081G
Vitamin K4 is a chemically synthesized Vitamin K that plays an important role in the normal blood coagulation system. It arrests cells in the S phase of the cell cycle and induces apoptosis, and can be used for research on cancer, including prostate cancer and osteosarcoma.
  • Inhibits cell proliferation and induces caspase-dependent apoptosis in PC-3 cells.
  • Induces S phase cell cycle arrest in PC-3 and U2OS cells.
  • Inhibits the growth of U2OS cells.
  • Increases ROS generation and disrupts mitochondrial membrane potential in U2OS cells.
  • Inhibits the migration of U2OS cells in vitro.
  • Modulates expression of Bcl-2, Bax, PARP, and cleaved-caspase-3.