Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin K4 98.0+%, TCI America™
SDP

Catalog No. V010325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V010325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V010325G Supplier TCI America Supplier No. V010325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 573-20-6
Molecular Formula C15H14O4
Molecular Weight (g/mol) 258.273
MDL Number MFCD00021469
InChI Key RYWSYCQQUDFMAU-UHFFFAOYSA-N
Synonym 1,4-Diacetoxy-2-methylnaphthalene, Menadiol Diacetate
PubChem CID 11310
IUPAC Name (4-acetyloxy-3-methylnaphthalen-1-yl) acetate
SMILES CC1=C(C2=CC=CC=C2C(=C1)OC(=O)C)OC(=O)C

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 25 g
Formula Weight 258.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Vitamin K4
EINECSNumber (9)-0040
RTECSNumber QJ5250000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.