Learn More
Medchemexpress LLC Vitamin K5 hydrochloride | 130-24-5 | 50 MG
SDP
Catalog No. 5000421037
Encompass_Preferred
Vitamin K5 hydrochloride acts as a photosensitizer and antimicrobial agent, specifically inhibiting PKM2 and inducing apoptosis in colon 26 cells. It finds use in research concerning infection and cancer, and as a preservative for various products.
  • Acts as a photosensitizer and antimicrobial agent
  • Functions as a specific PKM2 inhibitor
  • Induces apoptosis in colon 26 cells
  • Can be used for infection and cancer research
  • Suitable as a preservative for pharmaceuticals, foods, and beverages
  • Increases antibacterial effects of UVA irradiation on various bacteria (e.g., *Escherichia coli*, *Pseudomonas aeruginosa*, *Staphylococcus aureus*)
  • Generates superoxide anion radical and hydroxyl radical
  • Dose-dependently inhibits cellular glucose consumption and lactate production
  • Suppresses proliferation of colon 26 cells
  • Increases caspase-3 activity in colon 26 cells
Catalog No. 50-004-21037 Supplier Medchemexpress LLC Supplier No. HYN255250MG
Vitamin K5 hydrochloride acts as a photosensitizer and antimicrobial agent, specifically inhibiting PKM2 and inducing apoptosis in colon 26 cells. It finds use in research concerning infection and cancer, and as a preservative for various products.
  • Acts as a photosensitizer and antimicrobial agent
  • Functions as a specific PKM2 inhibitor
  • Induces apoptosis in colon 26 cells
  • Can be used for infection and cancer research
  • Suitable as a preservative for pharmaceuticals, foods, and beverages
  • Increases antibacterial effects of UVA irradiation on various bacteria (e.g., *Escherichia coli*, *Pseudomonas aeruginosa*, *Staphylococcus aureus*)
  • Generates superoxide anion radical and hydroxyl radical
  • Dose-dependently inhibits cellular glucose consumption and lactate production
  • Suppresses proliferation of colon 26 cells
  • Increases caspase-3 activity in colon 26 cells