Learn More
Medchemexpress LLC Vitamin u chloride (S-Methylmethionine sulfonium chloride) | 1115-84-0 | 25 MG
SDP
Catalog No. 5000420668
Encompass_Preferred
Vitamin U chloride is an orally active compound with anti-ulcer and antioxidant properties. It has been shown to inhibit adipocyte differentiation, promote skin wound healing, and is utilized in research concerning gastrointestinal ulceration.
  • Orally active anti-ulcer agent.
  • Possesses antioxidant activity.
  • Inhibits adipocyte differentiation.
  • Promotes skin wound healing.
  • Applicable in gastrointestinal ulceration research.
Catalog No. 50-004-20668 Supplier Medchemexpress LLC Supplier No. 25MGHYN2551
Vitamin U chloride is an orally active compound with anti-ulcer and antioxidant properties. It has been shown to inhibit adipocyte differentiation, promote skin wound healing, and is utilized in research concerning gastrointestinal ulceration.
  • Orally active anti-ulcer agent.
  • Possesses antioxidant activity.
  • Inhibits adipocyte differentiation.
  • Promotes skin wound healing.
  • Applicable in gastrointestinal ulceration research.