Learn More
Selleck Chemical LLC VitaMin U S4811-25mg
SDP
Catalog No. 503111540
Encompass_Preferred
Vitamin U (Methylmethioninesulfonium Chloride Cabagin-U Smethylmethionine) is a vitamin found in green vegetables It is used in the treatment of peptic ulcers colitis and gastritis and has an effect on secretory acid-forming and enzymatic functions of the intestinal tract
Catalog No. 50-311-1540 Supplier Selleck Chemical LLC Supplier No. S481125MG
Vitamin U (Methylmethioninesulfonium Chloride Cabagin-U Smethylmethionine) is a vitamin found in green vegetables It is used in the treatment of peptic ulcers colitis and gastritis and has an effect on secretory acid-forming and enzymatic functions of the intestinal tract