Description
Includes
Enterococcus faecalis ATCC™ 29212™, Enterococcus faecalis ATCC™ 51299™, Staphylococcus aureus subsp. aureus ATCC™ 29213™, Staphylococcus aureus subsp. aureus ATCC™ BAA-976™, Staphylococcus aureus subsp. aureus ATCC™ BAA-977™, Staphylococcus aureus ATCC™ BAA-1026™
- KWIK-STIK™
- Gives accurate, repeatable results
- Microorganism, hydrating fluid and inoculating swab all in one self-contained device
- Each device features single microorganism strain in lyophilized pellet, reservoir of hydrating fluid and inoculating swab
- For added convenience, peel-off identification label is included for easy documentation
- Available in packs of 2 or 6
Specifications
Specifications
| Includes | Enterococcus faecalis ATCC™ 29212™, Enterococcus faecalis ATCC™ 51299™, Staphylococcus aureus subsp. aureus ATCC™ 29213™, Staphylococcus aureus subsp. aureus ATCC™ BAA-976™, Staphylococcus aureus subsp. aureus ATCC™ BAA-977™, Staphylococcus aureus ATCC™ BAA-1026™ |
| Quantity | 2/Pk. |
| Specimen Type | Multiple |