Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Vitek™ 2 QC Sets
GSA_VA

Catalog No. R4652020 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
4 Loops/Set
5 Loops/Pk.
Product Type:
ANC Comp QC 1 Loop Set
ANC Comp QC Set
AST-GN QC 1 Loop Set
AST-GN QC Set
AST-GP QC 1 Loop Set
AST-GP QC Set
GN Comp QC 1 Loop Set
GN Comp QC Set
GP Comp QC 1 Loop Set
GP Comp QC Set
NH Comp QC Set
YST Comp QC 1 Loop Set
YST Comp QC Set
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
R4652020 5 Loops/Pk. GP Comp QC Set
R4652018 5 Loops/Pk. AST-GN QC Set
R4652021 5 Loops/Pk. GN Comp QC Set
R4652022 5 Loops/Pk. YST Comp QC Set
R4652019 5 Loops/Pk. AST-GP QC Set
R4652024 5 Loops/Pk. NH Comp QC Set
R4652030 5 Loops/Pk. ANC Comp QC Set
R4682021 5 Loops/Pk. GN Comp QC 1 Loop Set
R4682030 5 Loops/Pk. ANC Comp QC 1 Loop Set
22-246-508 4 Loops/Set AST-GN QC 1 Loop Set
22-246-509 5 Loops/Pk. AST-GP QC 1 Loop Set
R4682020 5 Loops/Pk. GP Comp QC 1 Loop Set
R4682022 5 Loops/Pk. YST Comp QC 1 Loop Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Catalog No. R4652020 Supplier Thermo Scientific™ Supplier No. R4652020
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Obtain faster recovery, enhanced viability, and direct-streak-to-plate capabilities with QC organisms manufactured using an unmatched proprietary gel preservation process.

Ensure high-performance products, reproducible clinical test results and consistent microbiological evaluation of pharmaceuticals and food with Thermo Scientific™ Culti-Loops™ quality control organisms. Thermo Scientific™ Vitek™ 2 QC Sets include a ready-to-use set of QC organisms in Culti-Loops format which are ready-to-use, disposable bacteriological loops containing stabilized, viable microorganisms in a unique gel matrix.

Trust Thermo Scientific quality control organisms for quality results you can depend on, backed by over 30 years of expertise in the manufacturing and packaging of controlled microorganisms.

Convenience

  • Decrease time and unnecessary costs of laboratory resources with a direct-streak inoculating loop that requires no rehydration.

Performance

  • Obtain enhanced viability and recovery with proprietary gel-preserved organisms.

Value

  • Consolidate ordering time and shipping costs by turning to one supplier for high-quality culture media and quality control organisms.

Confidence

  • Reach complete traceability with ATCC™ License Derivative strains.

Efficiency

  • Achieve accurate and convenient documentation with peel-off, transferable labels.

Safety

  • Reduce the risk of contamination and infection with zero organism handling.
  • More than 600 quality control organisms, including convenient sets for commonly used identification systems
  • ISO certified, cGMP (current Good Manufacturing Practices) compliant, FDA registered
  • ATCC™ Licensed Derivative Program member

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

*The ATCC Licensed Derivative™ Emblem, the ATCC Licensed Derivative™ word mark, and the ATCC catalog marks are trademarks of ATCC™. Thermo Fisher Scientific is licensed to use these trademarks and to sell products derived from ATCC™ cultures.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® BAA-750TM*, 700327™*, 49619™*, 43079™*, 43076™*, 29061™*, 19258™*, BAA-752™*
Certifications/Compliance ISO certified, cGMP (current Good Manufacturing Practices) compliant, FDA registered
Content And Storage 2°C to 8°C
Description Vitek 2 GP Comp QC Set/9
Format Culti-Loops
Product Type GP Comp QC Set
Target Organism Class Organism Sets
Test Type Qualitative QC
Quantity 5 Loops/Pk.
Species Listeria monocytogenes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.