Learn More
Cayman Chemical Vitellogenin Arctic char Po
SDP
Catalog No. 502968244
Encompass_Preferred
A nucleoside analog that competitively inhibits the equilibrative nucleoside transporter 1 (Kd 0.1-1.0 nM IC50s 4.6 and 3.6 nM in rat and human respectively) blocks adenosine uptake thereby potentiating the interaction of extracellular adenosine with purinoreceptors
Catalog No. 50-296-8244 Supplier Cayman Chemical Supplier No. 170120500 UL
A nucleoside analog that competitively inhibits the equilibrative nucleoside transporter 1 (Kd 0.1-1.0 nM IC50s 4.6 and 3.6 nM in rat and human respectively) blocks adenosine uptake thereby potentiating the interaction of extracellular adenosine with purinoreceptors