Learn More
Cayman Chemical Vitellogenin (fathead minnow) ELISA Kit, 96 Well
SDP
Catalog No. 502886974
Encompass_Preferred
The Vtg (fathead minnow) ELISA is a double-antibody immunometric (sandwich) ELISA for analyzing Vtg in whole body homogenates samples.
Catalog No. 50-288-6974 Supplier Cayman Chemical Supplier No. 1000694396 WELL
The Vtg (fathead minnow) ELISA is a double-antibody immunometric (sandwich) ELISA for analyzing Vtg in whole body homogenates samples.