Learn More
Cayman Chemical Vitellogenin rainbow trout E
SDP
Catalog No. 502886921
Encompass_Preferred
The Vtg (rainbow trout) ELISA is a double-antibody immunometric (sandwich) ELISA for analyzing Vtg in plasma samples.
Catalog No. 50-288-6921 Supplier Cayman Chemical Supplier No. 1000499496 WELL
The Vtg (rainbow trout) ELISA is a double-antibody immunometric (sandwich) ELISA for analyzing Vtg in plasma samples.