Learn More
Cayman Chemical Vitellogenin salmon Polyclo
SDP
Catalog No. 502965967
Encompass_Preferred
A form of the sugar Neu5Ac O-linked with the nucleotide CMP serves as the substrate for sialyltransferases which transfer Neu5Ac from CMP-sialic acid to various acceptor substrates most commonly at terminal positions of the oligosaccharide component of glycoproteins or glycolipids
Catalog No. 50-296-5967 Supplier Cayman Chemical Supplier No. 170140500 UL
A form of the sugar Neu5Ac O-linked with the nucleotide CMP serves as the substrate for sialyltransferases which transfer Neu5Ac from CMP-sialic acid to various acceptor substrates most commonly at terminal positions of the oligosaccharide component of glycoproteins or glycolipids