Learn More
Cayman Chemical Vitellogenin (salmonid) Semi-Quantitative ELISA Kit, 5 ea
SDP
Catalog No. 502886549
Encompass_Preferred
This assay is based on detection of Vtg by a monoclonal antibody, BN-5, which has high cross reactivity with Vtg from a variety of other species including Salmoniformes, Pleuronectiformes, and some Perciformes. This assay is a semi-quantitative assay for Vtg from fish plasma and it is not suitable for measuring absolute amounts of Vtg.
Catalog No. 50-288-6549 Supplier Cayman Chemical Supplier No. 100092725 EA
This assay is based on detection of Vtg by a monoclonal antibody, BN-5, which has high cross reactivity with Vtg from a variety of other species including Salmoniformes, Pleuronectiformes, and some Perciformes. This assay is a semi-quantitative assay for Vtg from fish plasma and it is not suitable for measuring absolute amounts of Vtg.