Learn More
Cayman Chemical Vitellogenin (carp) Standard, 1 ea
SDP
Catalog No. 502943957
Encompass_Preferred
The lyophilized carp Vtg may be used as a positive control in immunoblotting. Freshly reconstituted Vtg may also be used as a standard in a quantitative ELISA. This product contains bovine serum albumin which will appear as a strong protein band in acrylamide gels.
Catalog No. 50-294-3957 Supplier Cayman Chemical Supplier No. 4701141 EA
The lyophilized carp Vtg may be used as a positive control in immunoblotting. Freshly reconstituted Vtg may also be used as a standard in a quantitative ELISA. This product contains bovine serum albumin which will appear as a strong protein band in acrylamide gels.