Learn More
Cayman Chemical VItelogenIncod PolycLNal 500uL
SDP
Supplier:  Cayman Chemical 170130500 UL
Encompass_Preferred
Isoprostanesprostaglandin PG-like productsof freeradicalinducedlipidperoxidation.{320 Although the isoprostanes derived from arachidonic acid are the best characterized many other polyunsaturated fatty acids can form isoprostanes.{7323} (±)5-iPF2a-VI is one of dozens of possible stereo- and regioisomeric isoprostanes which can be formed from arachidonic acid. To date the most extensively studied of these is 8-isoprostane (8-epi-PGF2a iPF2a-III).{30645011} However 8-isoprostane is a minor isoprostane constituent when compared to some of the other isomers which form in natural conditions of oxidative stress.{8367} (±)5-iPF2a-VI is an isoprostane from the unique Type VI class of isoprostanes. This class has been shown to be one of the major isoprostane products in contrast to 8-isoprostane. In addition to being produced in greater abundance than 8-isoprostane Type VI isoprostanes form internal lactones which facilitates their extraction purification fro biological samples8367940869106390
Catalog No. 50-296-8443