Learn More
Cayman Chemical VItelogenInkIlIfIsh Mon 500uL
SDP
Catalog No. 502968123
Encompass_Preferred
An agonist of the group III metabotropic glutamate receptors mGluR4a and mGluR6 (EC50s 2-5 UM) mimics the phosphatidylserine head group and has been shown to inhibit the proliferation of microglia and to enhance neuronal differentiation of progenitor cells
Catalog No. 50-296-8123 Supplier Cayman Chemical Supplier No. 170195500 UL
An agonist of the group III metabotropic glutamate receptors mGluR4a and mGluR6 (EC50s 2-5 UM) mimics the phosphatidylserine head group and has been shown to inhibit the proliferation of microglia and to enhance neuronal differentiation of progenitor cells