Learn More
Cayman Chemical VItelogenInraInbow trout 10ug
SDP
Catalog No. 502945442
Encompass_Preferred
The lyophilized rainbow trout Vtgmay be used as a positive control in immunoblotting. Freshly reconstituted Vtg may also be used as a standard in a quantitative ELISA. The product contains bovine serum albumin which will appear as a strong protein band in acrylamide gels.
Catalog No. 50-294-5442 Supplier Cayman Chemical Supplier No. 47016410 UG
The lyophilized rainbow trout Vtgmay be used as a positive control in immunoblotting. Freshly reconstituted Vtg may also be used as a standard in a quantitative ELISA. The product contains bovine serum albumin which will appear as a strong protein band in acrylamide gels.