Learn More
Cayman Chemical VItelogenInsalmon Monclo 500uL
SDP
Catalog No. 502965596
Encompass_Preferred
A selective brain-penetrable LRRK2 inhibitor (Kis 2 and 18.7 nM in biochemical and cell-based assays respectively) inhibits LRRK2 Ser1292 autophosphorylation in BAC transgenic mice expressing human LRRK2 protein with the G2019S Parkinsons disease mutation (IC50 20 nM in vivo)
Catalog No. 50-296-5596 Supplier Cayman Chemical Supplier No. 170147500 UL
A selective brain-penetrable LRRK2 inhibitor (Kis 2 and 18.7 nM in biochemical and cell-based assays respectively) inhibits LRRK2 Ser1292 autophosphorylation in BAC transgenic mice expressing human LRRK2 protein with the G2019S Parkinsons disease mutation (IC50 20 nM in vivo)