Learn More
Cayman Chemical VItelogenInsalmon Monclo 500uL
SDP
Supplier:  Cayman Chemical 170145500 UL
Encompass_Preferred
A selective brain-penetrable LRRK2 inhibitor (Ki 0.7 nM) inhibits LRRK2 Ser1292 autophosphorylation in BAC transgenic mice expressing human LRRK2 protein with the G2019S Parkinsons disease mutation (IC50 3 nM in vivo)
Catalog No. 50-296-5327