Learn More
Glass Expansion Vitex Gripping Paper (PKT 6)
SDP
Catalog No. 502702973
Encompass_Preferred
Vitex Gripping Paper (PKT 6). Facilitates the insertion of capillary tubing into flared and non-flared peristaltic pump tubing to ensure a tight and secure fit that won’t disconnect. The gripping paper is folded over the end of the capillary tubing to maintain a tight grip on the tube while being inserted into the pump tubing. This gripping paper is especially handy while wearing PPE that can limit dexterity.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2973 Supplier Glass Expansion Supplier No. KK711E
Vitex Gripping Paper (PKT 6). Facilitates the insertion of capillary tubing into flared and non-flared peristaltic pump tubing to ensure a tight and secure fit that won’t disconnect. The gripping paper is folded over the end of the capillary tubing to maintain a tight grip on the tube while being inserted into the pump tubing. This gripping paper is especially handy while wearing PPE that can limit dexterity.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001