Learn More
GFS Chemicals Inc VITEX, (Modified Starch) | 100 G | GFS CHEMICALS, INC. Item #: 76601 | Bulk #: 331
SDP
Catalog No. NC0109345
Encompass_Preferred
VITEX, (Modified Starch) | Chemical Formula : | CAS # : 9005-84-9 | Specifications : Insoluble 0.01 % , Suitablilty as iodometric indicator
Catalog No. NC0109345 Supplier GFS Chemicals Inc Supplier No. 76601
VITEX, (Modified Starch) | Chemical Formula : | CAS # : 9005-84-9 | Specifications : Insoluble 0.01 % , Suitablilty as iodometric indicator