Learn More
AdipoGen Vitexin
SDP
Supplier:  AdipoGen AGCN20425M025
Encompass_Preferred
Chemical. CAS 3681-93-4. Formula C21H20O10. MW 432.4. Isolated from Crataegus pinnatifida. alpha-Glucosidase inhibitor. Antioxidant. HIF-1 alpha inhibitor. Anti-metastatic. Apoptosis inducer. Tumor suppressor. Adipogenesis inhibitor. Inhibits SIRT6 in vitro. Inhibits polyubiquitin synthesis by the ubiquitin-conjugating enzyme E2-25K. Cardioprotective. Anti-inflammatory. Shows analgesic effect by targeting TRPV1 channel activity. Neuroprotective. Inhibits NMDA receptors.
Catalog No. 50-256-2535