Learn More
Cayman Chemical VItexIn 2-O-rhamnosIde 10mg
SDP
Catalog No. 502934197
Encompass_Preferred
A flavonoid glycoside; inhibits DNA synthesis in MCF-7 breast cancer cells (IC50 = 17.5 μM)
Catalog No. 50-293-4197 Supplier Cayman Chemical Supplier No. 2681610 MG
A flavonoid glycoside; inhibits DNA synthesis in MCF-7 breast cancer cells (IC50 = 17.5 μM)