Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitexin 2-O-rhamnoside, ≥98.0% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111015066 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5066 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5066 Supplier MilliporeSigma Supelco Supplier No. 5560825MGF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 64820-99-1
Physical Form Neat
Linear Formula C27H30O14
MDL Number MFCD00143627
Synonym 2-O-Rhamnosylvitexin
Type Analytical Standard
Quantity 25 mg
Formula Weight 578.52
Percent Purity ≥98.0% (TLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.