Learn More
Medchemexpress LLC Vitexin 7-O-beta-D-glucopyranoside | 35109-95-6 | 594.52 | 1 MG
SDP
Catalog No. 5000416427
Encompass_Preferred
Vitexin 7-O-β-D-glucopyranoside (Vitexin 7-glucoside) is a flavone glycoside found in *Beta vulgaris* var. *cicla*. It shows hepatoprotective activity in primary cultured rat hepatocytes with Carbon tetrachloride (CCl4)-induced cell toxicity. This product is for research use only.
  • Found in *Beta vulgaris* var. *cicla*.
  • Demonstrates hepatoprotective activity.
  • For research use only.
  • Available as a light yellow to yellow solid.
  • Purity of 96.28%.
  • Stable as powder at -20°C for 3 years, 4°C for 2 years.
  • Stable in solvent at -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-004-16427 Supplier Medchemexpress LLC Supplier No. HYN151551MG
Vitexin 7-O-β-D-glucopyranoside (Vitexin 7-glucoside) is a flavone glycoside found in *Beta vulgaris* var. *cicla*. It shows hepatoprotective activity in primary cultured rat hepatocytes with Carbon tetrachloride (CCl4)-induced cell toxicity. This product is for research use only.
  • Found in *Beta vulgaris* var. *cicla*.
  • Demonstrates hepatoprotective activity.
  • For research use only.
  • Available as a light yellow to yellow solid.
  • Purity of 96.28%.
  • Stable as powder at -20°C for 3 years, 4°C for 2 years.
  • Stable in solvent at -80°C for 6 months, -20°C for 1 month.