Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitexin, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014050 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4050 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4050 Supplier MilliporeSigma Supelco Supplier No. 4951310MGF
Add to Cart
Add to Cart

Vitexin is a naturally derived C-glycosylated flavone found in various medicinal plants which can exhibit a wide range of anti-tumor, anti-oxidative and anti-inflammatory properties.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 3681-93-4
Physical Form Neat
Linear Formula C21H20O10
MDL Number MFCD00017456
Synonym 4 ′,5,7-Trihydroxyflavone 8-C-glucoside; 8-C-Glucosylapigenin; Apigenin 8-C-glucoside; Orientoside
Type Analytical Standard
Quantity 10 mg
Formula Weight 432.38
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.