Learn More
Abcam Vite x in (Apigenin 8-C-glucoside), Antio x idant agent, 25MG
Catalog No. 502705468
Encompass_Preferred
MW 432.4 Da, Purity >98%. Antioxidant agent. Potent glucosidase inhibitor (IC₅₀ = 48 nM). HIF-1 α inhibitor. Inhibits TNF-α, IL-1β, IL-6, and IL-33 production. Shows antinociceptive, anti-inflammatory, and antioxidant effects in vivo..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5468 Supplier Abcam Supplier No. AB14486225MG
MW 432.4 Da, Purity >98%. Antioxidant agent. Potent glucosidase inhibitor (IC₅₀ = 48 nM). HIF-1 α inhibitor. Inhibits TNF-α, IL-1β, IL-6, and IL-33 production. Shows antinociceptive, anti-inflammatory, and antioxidant effects in vivo..
The product is subject to the following: Abcam Restricted Use Statement