Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ VITLAB™ Tamper-Evident Closures

Catalog No. 14380105 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Thread Size (Metric):
GL 45
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
14-380-105 GL 45
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-380-105 Supplier BRANDTECH™ VITLAB™ Supplier No. 83330
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tamper-evident closures for VITgrip™ laboratory bottles.

  • Replacement closure
  • Polypropylene
  • GL 45 thread
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Description VITGRIP replacement screw caps with tamper evident closure, GL45, pack of 6
For Use With (Application) VITgrip bottles
Material Polypropylene
Quantity 6
Sterility Non-sterile
Thread Size (Metric) GL 45
Neck Finish 83 mm
Autoclavable Autoclavable
Disposable Single-use
Product Type Tamper-evident Cap
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.