Learn More
Sigma Aldrich Fine Chemicals Biosciences VitojectTM fluorocarbon ru
Supplier: Sigma Aldrich Fine Chemicals Biosciences Z6777361PR
Vitoject is made of 100% high chemical resistant Viton(R) absolutely indispensable when working with aromatic hydrocarbons (Benzene Toluene ...) chlorinated hydrocarbons (Tri Per ...) fuels aggressive acids and many other chemicals.Manufactured with the ecologically beneficial and modern Vulcoject-process technology (injection molding).Very good protection against extremely aggressive chemicals100% gastightExcellent wearing comfortSolvent free by eco-friendly injection molding processCE compliant