Learn More
Ace Glass, Inc. VITON 210 O-RING SEAL 6/PK VITON 210 O-RING SEAL 6/PK
SDP
Catalog No. NC0526519
Encompass_Preferred
Size 210 Viton o-ring. Pack/6. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. NC0526519 Supplier Ace Glass, Inc. Supplier No. 785530
Size 210 Viton o-ring. Pack/6. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.