Learn More
Analytical Sales & Services Viton Chemical-resistant External O-rings
Catalog No. 5000805102
Encompass
Viton Chemical-resistant External O-rings\nfor use with Flow Electrolysis Cells\n\n• Outer diameter: 5mm\n• Inner diameter: 3mm\n\nQty/Pkg: 50
Catalog No. 50-008-05102 Supplier Analytical Sales & Services Supplier No. F1088302
Viton Chemical-resistant External O-rings\nfor use with Flow Electrolysis Cells\n\n• Outer diameter: 5mm\n• Inner diameter: 3mm\n\nQty/Pkg: 50