Learn More
AVANTOR MASTERFLEX (VWR LLC) Viton FDA Compliant Tubing, 1/2"" (12.7 mm) ID
Catalog No. NC1695612
Encompass
Applications: FDA compliant food or laboratory applications, acid and solvent transfer, high temperature work. Characteristics: Good chemical resistance, opaque white. Certification: FDA compliant. Temperature range: -25 to 400¿F(-32 to 204¿C) Sterilize: Circulating hot air oven at 480¿F (249¿C)
Catalog No. NC1695612 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0643507
Applications: FDA compliant food or laboratory applications, acid and solvent transfer, high temperature work. Characteristics: Good chemical resistance, opaque white. Certification: FDA compliant. Temperature range: -25 to 400¿F(-32 to 204¿C) Sterilize: Circulating hot air oven at 480¿F (249¿C)