Learn More
Chemglass Life Sciences O-Ring, Viton, 100mm Flange
SDP
Catalog No. CG14721
Encompass_Preferred
O-Ring, Viton, 100mm Flange. O-ring for Chemglass reaction vessels and lids. Available in Viton, Silicone, Perfluoro, FEP encapsulated Silicone and FEP encapsulated Stainless Steel spring. The Stainless Steel spring maintains resilience even in long term exposure to cryogenic temperatures.Temperature Range:Viton: -30 to 200CSilicone: -50 to 230CPerfluoro: -7 to 230CFEP Encapsulated Silicone: -62 to 205CFEP Encapsulated Stainless: -250 to 205C
Catalog No. CG14721 Supplier Chemglass Life Sciences Supplier No. CG14721
O-Ring, Viton, 100mm Flange. O-ring for Chemglass reaction vessels and lids. Available in Viton, Silicone, Perfluoro, FEP encapsulated Silicone and FEP encapsulated Stainless Steel spring. The Stainless Steel spring maintains resilience even in long term exposure to cryogenic temperatures.Temperature Range:Viton: -30 to 200CSilicone: -50 to 230CPerfluoro: -7 to 230CFEP Encapsulated Silicone: -62 to 205CFEP Encapsulated Stainless: -250 to 205C