Learn More
MSC Value Collection ZMSCV2X12 12mm ID x 16mm OD Viton O-Ring 2mm Thick, Round Cross Section, Shore 70 - 80, -15 to 400°F
Catalog No. NC1963177
Encompass
Value Collection ZMSCV2X12 12mm ID x 16mm OD Viton O-Ring 2mm Thick, Round Cross Section, Shore 70 - 80, -15 to 400°F
Catalog No. NC1963177 Supplier MSC Supplier No. 31958945
Value Collection ZMSCV2X12 12mm ID x 16mm OD Viton O-Ring 2mm Thick, Round Cross Section, Shore 70 - 80, -15 to 400°F