Learn More
Chemglass Life Sciences O-Ring, Viton, 120mm Flange
SDP
Catalog No. CG14722
Encompass_Preferred
O-Ring, Viton, 120mm Flange. O-ring for Chemglass reaction vessels and lids. Available in Viton, Silicone, Perfluoro, FEP encapsulated Silicone and FEP encapsulated Stainless Steel spring. The Stainless Steel spring maintains resilience even in long term exposure to cryogenic temperatures.Temperature Range:Viton: -30 to 200CSilicone: -50 to 230CPerfluoro: -7 to 230CFEP Encapsulated Silicone: -62 to 205CFEP Encapsulated Stainless: -250 to 205C
Catalog No. CG14722 Supplier Chemglass Life Sciences Supplier No. CG14722
O-Ring, Viton, 120mm Flange. O-ring for Chemglass reaction vessels and lids. Available in Viton, Silicone, Perfluoro, FEP encapsulated Silicone and FEP encapsulated Stainless Steel spring. The Stainless Steel spring maintains resilience even in long term exposure to cryogenic temperatures.Temperature Range:Viton: -30 to 200CSilicone: -50 to 230CPerfluoro: -7 to 230CFEP Encapsulated Silicone: -62 to 205CFEP Encapsulated Stainless: -250 to 205C