Learn More
Waters Corp Waters O-Ring, FKM, 16.4 mm x 1 mm
Supplier: Waters Corp 5712021
This replacement O-ring (16.4 mm × 1 mm) is typically used as a seal in legacy ESI and APcI probe designs.
- Used as a seal in legacy ESI and APcI probe designs.
- Found in the probes of 3100, SQD, ACQUITY TQD, and earlier Mass Spectrometers.
- Also used in earlier versions of Xevo and Synapt products before the introduction of Tool Free Probes.