Learn More
MSC Value Collection ZMSCV70005 7/64" ID x 15/64" OD Dash 005 Viton O-Ring 1/16" Thick, Round Cross Section, Shore 70 - 80, -15 to 400°F
Catalog No. NC1673188
Encompass
Value Collection ZMSCV70005 7/64" ID x 15/64" OD Dash 005 Viton O-Ring 1/16" Thick, Round Cross Section, Shore 70 - 80, -15 to 400°F
Catalog No. NC1673188 Supplier MSC Supplier No. 09265059
Value Collection ZMSCV70005 7/64" ID x 15/64" OD Dash 005 Viton O-Ring 1/16" Thick, Round Cross Section, Shore 70 - 80, -15 to 400°F