Learn More
Waters Corp Viton O-ring, Dash 018
Supplier:  Waters Corp P6510AS018
Encompass_Preferred
This O-ring is a Viton O-ring, Dash 018, and is specifically designed for sealing applications in aqueous (PEEK) inline filter holders.
  • Made from Viton
  • Dash 018 specification
  • Designed for use in aqueous (PEEK) inline filter holders
Catalog No. NC1957786