Learn More
Ace Glass, Inc. VITON O RING SIZE 006 VITON O RING SIZE 006
SDP
Catalog No. NC0739540
Encompass_Preferred
Size 006 Viton o-ring. Pack/12. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. NC0739540 Supplier Ace Glass, Inc. Supplier No. 785501
Size 006 Viton o-ring. Pack/12. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.