Learn More
Glass Expansion Viton O-Rings for FDT/SDT Ball Joints (PKT 10)
SDP
Catalog No. 502703016
Encompass_Preferred
Viton O-Rings for FDT/SDT Ball Joints (PKT 10). Compatible with Agilent ICP-MS Semi Demountable Torch Injector Adaptor 30-808-4129, Analytic Jena PlasmaQuant 9000 Series ICP-OES Ball Joint Adaptor 31-807-0018, Nu Instruments TOF-ICP-MS Vitesse Quartz Torch 30-807-0560, and Shimadzu ICPS 7500/8100 and ICPE 9000/9800 ICP-OES Semi Demountable Torch Injector Adaptor 30-808-4129. The O-Ring removal tool (50-100-1636) facilitates the removal and installation of O-rings. This tool is especially handy while wearing PPE that can limit dexterity.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3016 Supplier Glass Expansion Supplier No. 70V011
Viton O-Rings for FDT/SDT Ball Joints (PKT 10). Compatible with Agilent ICP-MS Semi Demountable Torch Injector Adaptor 30-808-4129, Analytic Jena PlasmaQuant 9000 Series ICP-OES Ball Joint Adaptor 31-807-0018, Nu Instruments TOF-ICP-MS Vitesse Quartz Torch 30-807-0560, and Shimadzu ICPS 7500/8100 and ICPE 9000/9800 ICP-OES Semi Demountable Torch Injector Adaptor 30-808-4129. The O-Ring removal tool (50-100-1636) facilitates the removal and installation of O-rings. This tool is especially handy while wearing PPE that can limit dexterity.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001