Learn More
Perkin Elmer US LLC Viton O-ring, 24.99 mm I.D.
Catalog No. 502113651
Encompass_Preferred
Three of these o-rings are included in the Viton Burner Head O-Ring Kit for PinAAcle (09902219) and the Viton Burner Head O-Ring Kit for AAnalyst (09902219).
Catalog No. 50-211-3651 Supplier Perkin Elmer US LLC Supplier No. 09200079
Three of these o-rings are included in the Viton Burner Head O-Ring Kit for PinAAcle (09902219) and the Viton Burner Head O-Ring Kit for AAnalyst (09902219).