Learn More
Perkin Elmer US LLC Viton O-Ring for Gas Lines (Neb., Aux., Fuel) of End Cap
Catalog No. 502117303
Encompass_Preferred
The O-Ring is compatible with aqueous solutions and has dimensions of 0.312 I.D. x 0.070 W.D. Four O-rings are required for each end cap
Catalog No. 50-211-7303 Supplier Perkin Elmer US LLC Supplier No. 09200253
The O-Ring is compatible with aqueous solutions and has dimensions of 0.312 I.D. x 0.070 W.D. Four O-rings are required for each end cap