Learn More
Perkin Elmer US LLC Viton O-Ring for Nebulizers, 5.23 mm I.D.
Catalog No. 502117221
Encompass_Preferred
This o-ring has a 5.23 mm I.D. and 2.61 mm width. It is designed for use in MEINHARD nebulizers.
Catalog No. 50-211-7221 Supplier Perkin Elmer US LLC Supplier No. 00473194
This o-ring has a 5.23 mm I.D. and 2.61 mm width. It is designed for use in MEINHARD nebulizers.