Learn More
Waters Corp Waters O-Ring, Sample Cone, FKM, 12 x 1.5
Catalog No. NC1600419 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This O-ring is a replacement component designed for sample cones in various mass spectrometry systems, ensuring proper sealing and functionality.
  • Replacement sample cone O-ring.
  • Made from FKM material.
  • Features 12 x 1.5 dimensions.
  • Packaged as 1 unit per package.
  • Designed for MS system types.
  • Intended for replacement as required.
  • Compatible with source assemblies of 3100, SQD, SQD2, ACQUITY TQD, Xevo TQD, Xevo TQ-S, Xevo TQ-S Micro, Xevo TQ-S cronos, Xevo TQ-XS, Xevo TQ Absolute, Xevo G2-XS, Vion, SYNAPT G2-Si, SYNAPT XS, Select Series Cyclic IMS & MRT instruments.
Catalog No. NC1600419 Supplier Waters Corp Supplier No. 700001194
This O-ring is a replacement component designed for sample cones in various mass spectrometry systems, ensuring proper sealing and functionality.
  • Replacement sample cone O-ring.
  • Made from FKM material.
  • Features 12 x 1.5 dimensions.
  • Packaged as 1 unit per package.
  • Designed for MS system types.
  • Intended for replacement as required.
  • Compatible with source assemblies of 3100, SQD, SQD2, ACQUITY TQD, Xevo TQD, Xevo TQ-S, Xevo TQ-S Micro, Xevo TQ-S cronos, Xevo TQ-XS, Xevo TQ Absolute, Xevo G2-XS, Vion, SYNAPT G2-Si, SYNAPT XS, Select Series Cyclic IMS & MRT instruments.