Learn More
Glass Expansion Viton Pump Tube 2tag , 127mm spacing, 0.64mm ID, Orange/White (PKT 12)
SDP
Supplier:  Glass Expansion VS0.64OW
Encompass_Preferred
Viton Pump Tube 2tag, 127mm spacing, 0.64mm ID Orange/White (PKT 12).Tag Colors: Orange/WhiteID mm: 0.64Wall Thickness mm: 0.90OD mm 2.44Overall Length: 175mm (127mm spacing between tags)Number of Tubes Per Pack: 12Physical Properties: Fluoropolymer elastomerSpecial Properties: Special tubing for concentrated acids and corrosive solventsMaterial: Made from Fluran HC F-5500-A or equivalent.Limitations: Limited service lifeGlass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7728